siRNA Details
| Gene Symbol: Gpr89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026229.4 | NP_080505.1 | 9 | 863 | ||
| GenBank mRNA | AK009734.1 | - | 8 | 742 | ||
| AK016787.1 | - | 9 | 863 | |||
| AK034931.1 | - | 431 | ||||
| BC010729.1 | - | 9 | 856 | 3708143 | ||
More Information
Other Species Targeted
Set Membership: |
|

Gpr89
Genomic Map