siRNA Details
| Gene Symbol: Gstcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026231.2 | NP_080507.2 | 8 | 1651 | ||
| GenBank mRNA | AK013967.1 | - | 5 | 503 | ||
| AK017059.1 | - | 6 | 926 | |||
| AK020021.1 | - | 7 | 1261 | |||
| AK029724.1 | - | 8 | 1651 | |||
| AK033409.1 | - | 8 | 1689 | |||
| AK044441.1 | - | 8 | 1659 | |||
| BC047136.1 | - | 8 | 1651 | 6311866 | ||
| BC061240.1 | - | 8 | 1647 | 6743615 | ||

Gstcd
Genomic Map