siRNA Details
| Gene Symbol: SLC16A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006517.4 | NP_006508.2 | 1 | 546 | ||
| GenBank mRNA | AB085789.1 | - | 1 | 488 | ||
| CN342861.1 | - | 1 | 548 | |||
| U05315.1 | - | 1 | 578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC16A2| Gene Symbol: SLC16A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006517.4 | NP_006508.2 | 1 | 546 | ||
| GenBank mRNA | AB085789.1 | - | 1 | 488 | ||
| CN342861.1 | - | 1 | 548 | |||
| U05315.1 | - | 1 | 578 | |||