siRNA Details
| Gene Symbol: Mgat4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162368.1 | NP_001155840.1 | 5 | 568 | ||
| NM_001162369.1 | NP_001155841.1 | 5 | 441 | |||
| NM_001205098.1 | NP_001192027.1 | 7 | 981 | |||
| NM_026243.4 | NP_080519.2 | 3 | 463 | |||
| GenBank mRNA | AK018574.1 | - | 3 | 462 | ||
| AK018672.1 | - | 3 | 466 | |||
| AK032009.1 | - | 5 | 442 | |||
| AK033482.1 | - | 3 | 464 | |||
| BB611341.1 | - | 5 | 569 | |||
| BC046987.1 | - | 3 | 451 | 6475851 | ||

Mgat4c
Genomic Map