siRNA Details
| Gene Symbol: 4930430A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026248.3 | NP_080524.2 | 1 | 361 | ||
| GenBank mRNA | AK015247.1 | - | 1 | 360 | ||
| BC125324.1 | - | 1 | 312 | 40129839 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930430A15Rik| Gene Symbol: 4930430A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026248.3 | NP_080524.2 | 1 | 361 | ||
| GenBank mRNA | AK015247.1 | - | 1 | 360 | ||
| BC125324.1 | - | 1 | 312 | 40129839 | ||