siRNA Details
Gene Symbol: 4930524B15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026262.1 | NP_080538.1 | 3 | 427 | ||
GenBank mRNA | AK015006.1 | - | 3 | 469 | ||
AK015777.1 | - | 2 | 57 | |||
AK015879.1 | - | 3 | 427 | |||
BC114574.1 | - | 3 | 417 | 40049349 |
