siRNA Details
| Gene Symbol: 4930519G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026263.2 | NP_080539.1 | 12 | 1791 | ||
| GenBank mRNA | AK015843.1 | - | 12 | 1793 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930519G04Rik| Gene Symbol: 4930519G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026263.2 | NP_080539.1 | 12 | 1791 | ||
| GenBank mRNA | AK015843.1 | - | 12 | 1793 | ||