siRNA Details
| Gene Symbol: Fibin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026271.1 | NP_080547.1 | 1 | 1065 | ||
| GenBank mRNA | AK003790.1 | - | 1 | 1065 | ||
| AK011548.1 | - | 1 | 1236 | |||
| AK159648.1 | - | 1 | 1198 | |||
| BC027250.1 | - | 1 | 1054 | 3603588 | ||

Fibin
Genomic Map