siRNA Details
| Gene Symbol: Armc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026290.3 | NP_080566.2 | 5 | 691 | ||
| GenBank mRNA | AK005672.1 | - | 5 | 688 | ||
| AK015756.1 | - | 5 | 691 | |||
| BC049559.1 | - | 5 | 690 | 6707229 | ||
| EU477370.1 | - | 5 | 668 | |||

Armc12
Genomic Map