siRNA Details
| Gene Symbol: 4930542C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006987.1 | - | 624 | |||
| AK016017.1 | - | 682 | ||||
| AK017030.1 | - | 599 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930542C12Rik| Gene Symbol: 4930542C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006987.1 | - | 624 | |||
| AK016017.1 | - | 682 | ||||
| AK017030.1 | - | 599 | ||||