siRNA Details
| Gene Symbol: l7Rn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291286.1 | NP_001278215.1 | 2 | 435 | ||
| NM_001291287.1 | NP_001278216.1 | 3 | 499 | |||
| NM_001291288.1 | NP_001278217.1 | 2 | 435 | |||
| NM_026304.3 | NP_080580.1 | 2 | 435 | |||
| GenBank mRNA | AK002306.1 | - | 2 | 417 | ||
| AK012166.1 | - | 2 | 401 | |||
| AK206361.1 | - | 1 | 210 | |||
| BC003916.1 | - | 2 | 383 | 3485003 | ||
| BC083330.1 | - | 2 | 296 | |||
| DQ196315.1 | - | 2 | 413 | |||
| DQ196316.1 | - | 2 | 413 | |||

l7Rn6
Genomic Map