siRNA Details
| Gene Symbol: l7Rn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291286.1 | NP_001278215.1 | 5 | 982 | ||
| NM_001291287.1 | NP_001278216.1 | 6 | 1106 | |||
| NM_001291288.1 | NP_001278217.1 | 6 | 1183 | |||
| NM_001291289.1 | NP_001278218.1 | 4 | 804 | |||
| NM_026304.3 | NP_080580.1 | 5 | 1042 | |||
| GenBank mRNA | AK002306.1 | - | 5 | 1024 | ||

l7Rn6
Genomic Map