siRNA Details
| Gene Symbol: Pbdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026312.5 | NP_080588.1 | 6 | 1001 | ||
| GenBank mRNA | AK030507.1 | - | 6 | 983 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pbdc1| Gene Symbol: Pbdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026312.5 | NP_080588.1 | 6 | 1001 | ||
| GenBank mRNA | AK030507.1 | - | 6 | 983 | ||