siRNA Details
| Gene Symbol: Luc7l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026313.1 | NP_080589.1 | 10 | 1277 | ||
| GenBank mRNA | AK084714.1 | - | 5 | 664 | ||
| AK151228.1 | - | 10 | 1325 | |||
| AK151839.1 | - | 10 | 1325 | |||
| BC009092.1 | - | 10 | 1277 | 3157532 | ||

Luc7l3
Genomic Map