siRNA Details
| Gene Symbol: Kirrel3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190911.1 | NP_001177840.1 | 17 | 2496 | ||
| NM_001190912.1 | NP_001177841.1 | 16 | 2493 | |||
| NM_001190914.1 | NP_001177843.1 | 17 | 2561 | |||
| NM_026324.3 | NP_080600.1 | 16 | 2460 | |||
| GenBank mRNA | AK045373.1 | - | 17 | 2401 | ||
| AK079332.1 | - | 17 | 2499 | |||
| AK171866.1 | - | 13 | 1811 | |||
| AY169782.1 | - | 16 | 2460 | |||
| BC063072.1 | - | 16 | 2288 | 5695003 | ||

Kirrel3
Genomic Map