siRNA Details
| Gene Symbol: Reg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026328.2 | NP_080604.2 | 3 | 310 | ||
| GenBank mRNA | AK008049.1 | - | 3 | 312 | ||
| AK008438.1 | - | 3 | 313 | |||
| AK160329.1 | - | 3 | 307 | |||
| AK162316.1 | - | 3 | 311 | |||
| BC019465.1 | - | 3 | 293 | 4216795 | ||

Reg4
Genomic Map