siRNA Details
| Gene Symbol: Lipf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026334.3 | NP_080610.1 | 6 | 636 | ||
| GenBank mRNA | AK009300.1 | - | 6 | 638 | ||
| AK009359.1 | - | 6 | 638 | |||
| AK009413.1 | - | 6 | 637 | |||
| AK009428.1 | - | 6 | 640 | |||
| AK009431.1 | - | 6 | 640 | |||
| AK009437.1 | - | 6 | 638 | |||
| AK009459.1 | - | 6 | 638 | |||
| AK009473.1 | - | 6 | 637 | |||
| AK009474.1 | - | 6 | 638 | |||
| AK009479.1 | - | 6 | 635 | |||
| AK009523.1 | - | 6 | 638 | |||
| AK009525.1 | - | 6 | 638 | |||
| AK009537.1 | - | 6 | 638 | |||
| AK009544.1 | - | 6 | 633 | |||
| AK009546.1 | - | 6 | 641 | |||
| AK009560.1 | - | 6 | 638 | |||
| AK009571.1 | - | 6 | 637 | |||
| AK009573.1 | - | 6 | 641 | |||
| AK009729.1 | - | 6 | 638 | |||
| AK009773.1 | - | 6 | 638 | |||
| AK009875.1 | - | 6 | 638 | |||
| AK009923.1 | - | 6 | 699 | |||
| AK009990.1 | - | 6 | 638 | |||
| AK010019.1 | - | 6 | 639 | |||
| AK010026.1 | - | 6 | 638 | |||
| AK010035.1 | - | 6 | 638 | |||
| AK010036.1 | - | 6 | 637 | |||
| AK010058.1 | - | 6 | 637 | |||
| AK010061.1 | - | 6 | 638 | |||
| AK010091.1 | - | 6 | 636 | |||
| AK010093.1 | - | 6 | 638 | |||
| AK010103.1 | - | 6 | 639 | |||
| AK010106.1 | - | 6 | 638 | |||
| AK010116.1 | - | 6 | 636 | |||
| AK010123.1 | - | 6 | 639 | |||
| AK010124.1 | - | 5 | 632 | |||
| AK010125.1 | - | 6 | 638 | |||
| AK010139.1 | - | 6 | 638 | |||
| AK010203.1 | - | 6 | 639 | |||
| AK010231.1 | - | 6 | 637 | |||
| AK010236.1 | - | 6 | 639 | |||
| BC061067.1 | - | 6 | 636 | 30301084 | ||

Lipf
Genomic Map