siRNA Details
| Gene Symbol: Dph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026344.3 | NP_080620.1 | 6 | 1941 | ||
| GenBank mRNA | AK011199.1 | - | 6 | 1959 | ||
| AK018638.1 | - | 6 | 1942 | |||
| AK051026.1 | - | 6 | 1961 | |||
| AK080747.1 | - | 4 | 2238 | |||
| AK194858.1 | - | 1 | 50 | |||
| AK218933.1 | - | 1 | 78 | |||
| BC014746.1 | - | 5 | 1837 | |||
| BC031124.1 | - | 6 | 1928 | 5029876 | ||

Dph2
Genomic Map