siRNA Details
| Gene Symbol: Itgb3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026348.3 | NP_080624.1 | 4 | 248 | ||
| GenBank mRNA | AK005779.1 | - | 4 | 317 | ||
| AK013373.1 | - | 4 | 263 | |||
| AK015557.1 | - | 4 | 273 | |||
| AK045037.1 | - | 4 | 249 | |||
| AK166362.1 | - | 4 | 249 | |||
| BC049557.1 | - | 4 | 258 | 6741650 | ||
| EF437329.1 | - | 2 | 156 | |||

Itgb3bp
Genomic Map