siRNA Details
| Gene Symbol: Gpatch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026367.4 | NP_080643.4 | 2 | 208 | ||
| GenBank mRNA | AK011727.1 | - | 2 | 193 | ||
| AK017975.1 | - | 2 | 177 | |||
| AK029990.1 | - | 2 | 193 | |||
| AK030026.1 | - | 2 | 207 | |||
| AK032734.1 | - | 2 | 183 | |||
| AK053781.1 | - | 2 | 200 | |||
| AK167826.1 | - | 2 | 212 | |||
| AK169660.1 | - | 2 | 209 | |||
| BC054810.1 | - | 2 | 180 | 6839419 | ||

Gpatch2
Genomic Map