siRNA Details
| Gene Symbol: Gpatch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026367.4 | NP_080643.4 | 2 | 413 | ||
| GenBank mRNA | AK011727.1 | - | 2 | 398 | ||
| AK017975.1 | - | 2 | 382 | |||
| AK029990.1 | - | 2 | 398 | |||
| AK030026.1 | - | 2 | 412 | |||
| AK032734.1 | - | 2 | 388 | |||
| AK053781.1 | - | 2 | 405 | |||
| AK167826.1 | - | 2 | 417 | |||
| AK169660.1 | - | 2 | 414 | |||
| BC054810.1 | - | 2 | 385 | 6839419 | ||

Gpatch2
Genomic Map