siRNA Details
| Gene Symbol: Caap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026368.2 | NP_080644.2 | 6 | 1489 | ||
| GenBank mRNA | AK076487.1 | - | 6 | 1444 | ||
| AK140382.1 | - | 6 | 1491 | |||
| AK150311.1 | - | 7 | 1399 | |||
| AK160297.1 | - | 6 | 1492 | |||
| AK160572.1 | - | 6 | 1490 | |||
| AK171141.1 | - | 6 | 1491 | |||
| BC020067.1 | - | 6 | 1477 | 3991510 | ||

Caap1
Genomic Map