siRNA Details
| Gene Symbol: Loh12cr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170479.1 | NP_001163950.1 | 4 | 1434 | ||
| NM_026371.3 | NP_080647.2 | 4 | 1576 | |||
| GenBank mRNA | AK007423.1 | - | 4 | 1433 | ||
| AK018019.1 | - | 4 | 1492 | |||
| AK151342.1 | - | 4 | 1442 | |||
| AK169615.1 | - | 4 | 1433 | |||
| BC023059.1 | - | 4 | 1520 | 5355560 | ||

Loh12cr1
Genomic Map