siRNA Details
| Gene Symbol: Wdr83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026399.2 | NP_080675.2 | 1, 2 | 227 | ||
| GenBank mRNA | AK005790.1 | - | 1, 2 | 167 | ||
| AK020654.1 | - | 1, 2 | 188 | |||
| AY365434.1 | - | 1, 2 | 227 | |||
| BC005587.1 | - | 1, 2 | 184 | |||
| BC019369.1 | - | 1, 2 | 150 | 4014928 | ||
| BC020047.1 | - | 787 | ||||
| BC034842.1 | - | 1, 2 | 113 | |||
| BC094624.1 | - | 1, 2 | 164 | |||

Wdr83
Genomic Map