siRNA Details
| Gene Symbol: Wdr83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026399.2 | NP_080675.2 | 2 | 273 | ||
| GenBank mRNA | AK005790.1 | - | 2 | 213 | ||
| AK020654.1 | - | 2 | 234 | |||
| AY365434.1 | - | 2 | 273 | |||
| BC005587.1 | - | 2 | 230 | |||
| BC019369.1 | - | 2 | 196 | 4014928 | ||
| BC020047.1 | - | 833 | ||||
| BC034842.1 | - | 2 | 159 | |||
| BC094624.1 | - | 2 | 210 | |||

Wdr83
Genomic Map