siRNA Details
| Gene Symbol: Tmem39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205286.1 | NP_001192215.1 | 5 | 665 | ||
| NM_001205287.1 | NP_001192216.1 | 5 | 917 | |||
| NM_026407.3 | NP_080683.2 | 5 | 917 | |||
| GenBank mRNA | AK011646.1 | - | 5 | 891 | ||
| AK017817.1 | - | 5 | 875 | |||
| AK160576.1 | - | 5 | 666 | |||
| AK169127.1 | - | 5 | 919 | |||
| BC020318.2 | - | 5 | 872 | 5250532 | ||

Tmem39a
Genomic Map