siRNA Details
| Gene Symbol: Tmem39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205286.1 | NP_001192215.1 | 7 | 1221 | ||
| NM_001205287.1 | NP_001192216.1 | 7 | 1269 | |||
| NM_026407.3 | NP_080683.2 | 7 | 1473 | |||
| GenBank mRNA | AK011646.1 | - | 7 | 1450 | ||
| AK017817.1 | - | 7 | 1431 | |||
| AK133638.1 | - | 7 | 1445 | |||
| AK160576.1 | - | 7 | 1222 | |||
| AK169127.1 | - | 7 | 1271 | |||
| BC020318.2 | - | 7 | 1428 | 5250532 | ||

Tmem39a
Genomic Map