siRNA Details
| Gene Symbol: Yipf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026417.4 | NP_080693.2 | 6 | 1677 | ||
| GenBank mRNA | AK009618.1 | - | 6 | 1668 | ||
| AK017721.1 | - | 6 | 1653 | |||
| AK017722.1 | - | 6 | 1656 | |||
| AK083301.1 | - | 6 | 1657 | |||
| AK086213.1 | - | 382 | ||||
| AK159227.1 | - | 6 | 1639 | |||
| AK166347.1 | - | 6 | 1667 | |||
| BC003936.1 | - | 7 | 1852 | 3486503 | ||

Yipf4
Genomic Map