siRNA Details
| Gene Symbol: Uxs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026430.3 | NP_080706.1 | 6, 7 | 449 | ||
| GenBank mRNA | AF399958.1 | - | 6, 7 | 449 | ||
| AK005536.1 | - | 6, 7 | 389 | |||
| AK075806.1 | - | 6, 7 | 501 | |||
| AK076166.1 | - | 5, 6 | 490 | |||
| AK152376.1 | - | 6, 7 | 504 | |||
| AK208821.1 | - | 1, 2 | 188 | |||
| BC037049.1 | - | 6, 7 | 495 | 4935237 | ||

Uxs1
Genomic Map