siRNA Details
| Gene Symbol: Tmem86a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026436.3 | NP_080712.1 | 3 | 1639 | ||
| GenBank mRNA | AK007864.1 | - | 3 | 1640 | ||
| AK075589.1 | - | 3 | 1613 | |||
| BC056619.1 | - | 3 | 1606 | 4987281 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem86a| Gene Symbol: Tmem86a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026436.3 | NP_080712.1 | 3 | 1639 | ||
| GenBank mRNA | AK007864.1 | - | 3 | 1640 | ||
| AK075589.1 | - | 3 | 1613 | |||
| BC056619.1 | - | 3 | 1606 | 4987281 | ||