siRNA Details
| Gene Symbol: Zfp169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164575.1 | NP_001158047.1 | 5 | 685 | ||
| NM_026450.3 | NP_080726.3 | 5 | 717 | |||
| GenBank mRNA | BC060222.1 | - | 5 | 675 | 30356624 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp169| Gene Symbol: Zfp169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164575.1 | NP_001158047.1 | 5 | 685 | ||
| NM_026450.3 | NP_080726.3 | 5 | 717 | |||
| GenBank mRNA | BC060222.1 | - | 5 | 675 | 30356624 | |