siRNA Details
| Gene Symbol: Coq9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026452.2 | NP_080728.1 | 7 | 823 | ||
| GenBank mRNA | AK004527.1 | - | 3 | 248 | ||
| AK009179.1 | - | 7 | 835 | |||
| AK154343.1 | - | 7 | 848 | |||
| AK158979.1 | - | 7 | 842 | |||
| BC036386.1 | - | 7 | 823 | 1067021 | ||

Coq9
Genomic Map