siRNA Details
| Gene Symbol: Serpini2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026460.3 | NP_080736.2 | 2 | 218 | ||
| GenBank mRNA | AF251276.1 | - | 2 | 203 | ||
| AK007347.1 | - | 2 | 219 | |||
| AK007510.1 | - | 2 | 193 | |||
| AK008931.1 | - | 2 | 219 | |||
| BC098466.1 | - | 2 | 195 | 6433250 | ||

Serpini2
Genomic Map