siRNA Details
| Gene Symbol: 1700129C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026461.2 | NP_080737.1 | 5 | 988 | ||
| GenBank mRNA | AK007313.1 | - | 5 | 988 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700129C05Rik| Gene Symbol: 1700129C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026461.2 | NP_080737.1 | 5 | 988 | ||
| GenBank mRNA | AK007313.1 | - | 5 | 988 | ||