siRNA Details
Gene Symbol: SLU7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006425.4 | NP_006416.3 | 2 | 274 | ||
XM_005265793.3 | XP_005265850.1 | 2 | 621 | |||
XM_011534418.2 | XP_011532720.1 | 3 | 324 | |||
GenBank mRNA | AF101074.1 | - | 1 | 119 | ||
AK024875.1 | - | 2 | 194 | |||
AK314564.1 | - | 2 | 194 | |||
AY486334.1 | - | 2 | 154 | |||
BC010634.1 | - | 2 | 200 | 3871882 | ||
DA232975.1 | - | 2 | 276 | |||
DA409854.1 | - | 2 | 200 | |||
DQ174516.1 | - | 2 | 206 | |||
DQ174517.1 | - | 2 | 244 |
