siRNA Details
| Gene Symbol: Ccny | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026484.3 | NP_080760.2 | 1, 2 | 640 | ||
| GenBank mRNA | AK014201.1 | - | 1, 2 | 645 | ||
| AK017493.1 | - | 1, 2 | 472 | |||
| AK028696.1 | - | 1, 2 | 641 | |||
| AK076556.1 | - | 1, 2 | 170 | |||
| BC023321.1 | - | 2, 3 | 558 | 3995040 | ||

Ccny
Genomic Map