siRNA Details
| Gene Symbol: 1110004E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026502.2 | NP_080778.2 | 3, 4 | 497 | ||
| GenBank mRNA | AK003413.1 | - | 3, 4 | 388 | ||
| AK018510.1 | - | 3, 4 | 468 | |||
| AK045905.1 | - | 3, 4 | 476 | |||
| BC019533.1 | - | 3, 4 | 461 | 4191729 | ||
| CX236612.1 | - | 3, 4 | 497 | |||

1110004E09Rik
Genomic Map