siRNA Details
| Gene Symbol: Bambi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026505.2 | NP_080781.1 | 3 | 1822 | ||
| GenBank mRNA | AK045192.1 | - | 3 | 1822 | ||
| AK143185.1 | - | 3 | 1837 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bambi| Gene Symbol: Bambi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026505.2 | NP_080781.1 | 3 | 1822 | ||
| GenBank mRNA | AK045192.1 | - | 3 | 1822 | ||
| AK143185.1 | - | 3 | 1837 | |||