siRNA Details
| Gene Symbol: Apopt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026511.2 | NP_080787.1 | 4, 5 | 509 | ||
| GenBank mRNA | AK002873.1 | - | 4, 5 | 511 | ||
| AK005403.1 | - | 4, 5 | 497 | |||
| AK010493.1 | - | 4, 5 | 499 | |||
| AK012648.1 | - | 4, 5 | 507 | |||
| AK029408.1 | - | 4, 5 | 485 | |||
| BC071262.1 | - | 4, 5 | 509 | 6509633 | ||

Apopt1
Genomic Map