siRNA Details
| Gene Symbol: 2810417H13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026515.2 | NP_080791.2 | 4 | 1888 | ||
| GenBank mRNA | AK013105.1 | - | 4 | 1848 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810417H13Rik| Gene Symbol: 2810417H13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026515.2 | NP_080791.2 | 4 | 1888 | ||
| GenBank mRNA | AK013105.1 | - | 4 | 1848 | ||