siRNA Details
| Gene Symbol: Tmem178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026516.2 | NP_080792.2 | 3, 4 | 717 | ||
| GenBank mRNA | AK013110.1 | - | 3, 4 | 718 | ||
| AK014196.1 | - | 3, 4 | 717 | |||
| BX511370.1 | - | 3, 4 | 325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem178| Gene Symbol: Tmem178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026516.2 | NP_080792.2 | 3, 4 | 717 | ||
| GenBank mRNA | AK013110.1 | - | 3, 4 | 718 | ||
| AK014196.1 | - | 3, 4 | 717 | |||
| BX511370.1 | - | 3, 4 | 325 | |||