siRNA Details
| Gene Symbol: Fam173b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026546.3 | NP_080822.1 | 5 | 1041 | ||
| GenBank mRNA | AK003724.2 | - | 2 | 570 | ||
| AK020868.1 | - | 5 | 1041 | |||
| AK044518.1 | - | 568 | ||||
| AK160923.1 | - | 1168 | ||||
| BC030179.1 | - | 4 | 1032 | 5039149 | ||

Fam173b
Genomic Map