siRNA Details
| Gene Symbol: Yif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026553.4 | NP_080829.1 | 3 | 437 | ||
| GenBank mRNA | AK011071.1 | - | 3 | 431 | ||
| AK077329.1 | - | 3 | 423 | |||
| AK160409.1 | - | 3 | 390 | |||
| BC011117.1 | - | 3 | 379 | 4190535 | ||

Yif1a
Genomic Map