siRNA Details
| Gene Symbol: Apool | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026565.2 | NP_080841.1 | 3 | 237 | ||
| GenBank mRNA | AK012398.1 | - | 3 | 283 | ||
| AK012804.1 | - | 3 | 278 | |||
| AK028953.1 | - | 3 | 298 | |||
| AK079141.1 | - | 3 | 288 | |||
| BC024334.1 | - | 3 | 237 | 5036743 | ||

Apool
Genomic Map