siRNA Details
| Gene Symbol: Cgnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018850.1 | - | 3 | 651 | ||
| AK019857.1 | - | 3 | 638 | |||
| AK173252.1 | - | 17 | 2875 | |||
| BC031499.1 | - | 3 | 487 | |||
| BC039211.1 | - | 11 | 1593 | |||
| BC139006.1 | - | 18 | 4019 | 8862028 | ||
| BC158061.1 | - | 18 | 4022 | 9007361 | ||

Cgnl1
Genomic Map