siRNA Details
| Gene Symbol: Cgnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018850.1 | - | 3 | 895 | ||
| AK019857.1 | - | 3 | 882 | |||
| AK173252.1 | - | 17 | 3119 | |||
| BC031499.1 | - | 3 | 731 | |||
| BC039211.1 | - | 11 | 1837 | |||
| BC139006.1 | - | 18 | 4263 | 8862028 | ||
| BC158061.1 | - | 18 | 4266 | 9007361 | ||

Cgnl1
Genomic Map