siRNA Details
| Gene Symbol: Fam135a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026604.4 | NP_080880.4 | 7 | 837 | ||
| GenBank mRNA | AK053786.1 | - | 7 | 756 | ||
| AK081262.1 | - | 7 | 782 | |||
| AK149303.1 | - | 7 | 829 | |||
| AK165306.1 | - | 7 | 838 | |||
| BC070446.1 | - | 6 | 519 | 30362784 | ||

Fam135a
Genomic Map