siRNA Details
| Gene Symbol: Nudt21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026623.3 | NP_080899.1 | 4, 5 | 618 | ||
| GenBank mRNA | AK011688.2 | - | 4, 5 | 608 | ||
| AK012308.1 | - | 4, 5 | 592 | |||
| AK019433.1 | - | 4, 5 | 601 | |||
| AK020150.1 | - | 4, 5 | 599 | |||
| AK146419.1 | - | 4, 5 | 584 | |||
| AK147061.1 | - | 601 | ||||
| AK160147.1 | - | 4, 5 | 584 | |||
| AK208594.1 | - | 3, 4 | 193 | |||
| BC008270.1 | - | 4, 5 | 601 | 2811199 | ||
| BC090834.1 | - | 577 | 6855233 | |||

Nudt21
Genomic Map