siRNA Details
| Gene Symbol: Mturn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289740.1 | NP_001276669.1 | 3 | 544 | ||
| GenBank mRNA | AK019120.1 | - | 3 | 425 | ||
| AK019128.1 | - | 3 | 423 | |||
| AK043452.1 | - | 3 | 397 | |||
| BC042507.1 | - | 3 | 436 | 4235651 | ||
| BC052467.1 | - | 3 | 428 | 6834900 | ||

Mturn
Genomic Map