siRNA Details
| Gene Symbol: A930018P22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026634.2 | NP_080910.1 | 2 | 689 | ||
| GenBank mRNA | AK006944.1 | - | 2 | 683 | ||
| AK020874.1 | - | 2 | 690 | |||
| BC048438.1 | - | 2 | 648 | 6706651 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930018P22Rik| Gene Symbol: A930018P22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026634.2 | NP_080910.1 | 2 | 689 | ||
| GenBank mRNA | AK006944.1 | - | 2 | 683 | ||
| AK020874.1 | - | 2 | 690 | |||
| BC048438.1 | - | 2 | 648 | 6706651 | ||